Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ortho-Clinical Diagnostics™ VITROS™ Calibrators: Free T4
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Ortho-Clinical Diagnostics™ 1728872
This item is not returnable.
View return policy
Specifications
2°C to 8°C | |
Human Serum | |
VITROS™ Free T4 calibrators 1, 2 and 3 (freeze-dried, T4 in Human Serum with antimicrobial agent, reconstitution volume 1mL), lot calibration card, protocol card, 24 calibrator bar code labels (8 for each calibrator) | |
3 | |
3 Sets/Pk. |
VITROS™ ECi/ECiQ immunodiagnostic system, 3600 immunodiagnostic system, 5600 integrated system | |
Free T4 Calibrator | |
Quantitative measurement of Free Thyroxine (FT4) | |
Yes |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction