Learn More
Thermo Scientific™ Orion™ AQUAfast™ AQ3170 Colorimeter
Gather accurate chlorine measurements with a colorimeter designed for both the laboratory and the field in markets such as drinking water, wastewater, food & beverage, aquaculture, and agriculture.
Supplier: Thermo Scientific™ AC2V10
Description
The Thermo Scientific™ Orion™ AQUAfast™ AQ3170 colorimeter has a rugged and waterproof design to provide accurate chlorine measurements whether testing in the laboratory or the field. This colorimeter is designed to improve the user experience by including a large, backlit display, a real time clock and automatic data logging. The AQUAfast AQ3170 colorimeter is used in markets such as drinking water, wastewater, food & beverage, aquaculture, and agriculture.
- Large digital backlit display
- Automatic switch-off
- User friendly keypad
- Real time clock and date
- Calibration mode
- Storage function
- Waterproof sample chamber
- Angled, ergonomic design for easy bench top or hand-held use
- Suppression of chloride interference
- Ability to export data to a PC
- ST-COD tubes ready for use
- 2-year warranty
Warranty and Services
2 years
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
AQUAfast 10mm short path-length plastic vials for high level chlorine, 12 pack | |
AQ3170 Chlorine Colorimeter | |
12 pack of 10 mm plastic short path-length vials |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.