Learn More
Thermo Scientific™ Orion™ 927006MD Glass ATC Probe
Automatically correct pH measurements for variation in electrode slope due to temperature change with this glass ATC probe.
Supplier:  Thermo Scientific™ 927006
 View more versions of this product
Catalog No. 13-301-138
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
Automatically correct pH measurements for variation in electrode slope due to temperature change with the Thermo Scientific™ Orion™ 927006MD Glass ATC Probe. Available with three different connector options.
Note:
Three different models are available to accommodate different Orion meters. MiniDIN version is compatible with older meters with use of an adapter cable.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0°C to 100°C
±1°C
0 to 14
3.5 mm Phono Jack connector (ATC)
8 mm
120 mm
3.5 mm Phono Jack
Glass
ATC Probe, Glass
Orion Star Series Meters
0.3 in.
4.7 in.
32°F to 212°F
Store in air
Product Suggestions
Product Suggestions