Learn More
Dentec Safety™ Organic Vapor Particulate Cartridge
Dentec Safety™ Organic Vapor Particulate Cartridge features unique space plenums that allow users to access the entire filter material reducing breathing resistance, providing longer cartrdige life, reducing costs and improving productivity.
$43.40 - $687.00
Specifications
Material | Plastic |
---|---|
Product Type | Respirator Accessories |
For Use With (Equipment) | Masks |
Includes | Cartridges and filters |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
19-222-507
|
Dentec Safety™
158T20 |
6/Box |
Pack of 6 for $43.40
|
|
|||||
19-222-508
|
Dentec Safety™
158T20B100 |
100/Box |
Case of 100 for $687.00
|
|
|||||
Description
- Offset connector allows the user to rotate the cartridge into 3 different positions to improve visibility and allow faceshields or welding helmets to be worn without interference
- Saf-Twist design quickly attaches the cartridge to any Dentec Safety half-mask or full face air-purifying respirator Facepiece.
Specifications
Plastic | |
Respirator Accessories | |
Masks | |
Cartridges and filters |