Learn More
AGI NORTH AMERICA INC Syrris Orb Pilot floor-standing jacketed reactor for scale-up processes, available from 10L to 50L
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: AGI NORTH AMERICA INC 30002OPBF
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
![Request Bulk Order](/content/dam/fishersci/glyphs/15395_Quote_Bulk_EN.jpg)
Orb Pilot has multiple setups in one footprint.Included:-10L jacketed glass vessel (10 - 50L available separately)-Optimised jacket inlet to ensure high-efficiency heat transfer-Detachable outlet valve for easy cleaning-Base frame adaptable to multiple vessel sizes-Spring-loaded vessel support for easy vessel changes-Force-limited vessel clamp to prevent breakage from over-tightening-Operating temperature range: -40 to 235 deg C-Pressure range: Vacuum: ≤50 mbara. Maximum pressure: 250 mbar-LEMO RTD probe-Oil drain valve allows for easy draining of vessel jackets -Frame-mounted pair of short oil pipes with connectors-PTFE anchor stirrer with stirrer seal & guide -Lid blanking kitOrder informationNot included: appropriate circulator and pair of oil pipes from the frame-mounted short oil pipes to the circulator. Oil pipes required M30 x 1.5 female connection and adapter depending on the circulator.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.