Learn More
Med Vet International OPHTHALMIC STRIPSROSE100BX
Supplier: Med Vet International JOR1191R
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Rose Bengal is a diagnostic agent used for staining dead degenerated epithelial cells ¿ Individually packaged in sterile chevron pouch¿ No anesthesia necessary¿ 100 strips per box
This item is not returnable.
View return policy