Learn More
Bellco Glass, Inc. Manufactured from type 1 class a Borosilicate glass. Includes 4 or 6 45mm polypropylene blue screw caps and pouring rings, also 1 100mm polypropylene blue cap for the center. Suitable for all cell culture fermentation. Compatible with bellco over head dri
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Bellco Glass, Inc. 196415000
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Manufactured from type 1 class a Borosilicate glass. Includes 4 or 6 45mm polypropylene blue screw caps and pouring rings, also 1 100mm polypropylene blue cap for the center. Suitable for all cell culture fermentation. Compatible with bellco over head dri