Learn More
Research Products International Corp OHAUS Reciprocating Shaker, SHRC0719DG
SDP
Supplier:  Research Products International Corp OH30391828
Encompass_Preferred
OHAUS Reciprocating Shakers are designed for a range of applications including cell cultures and extraction procedures that require accurate, repeatable results. Our shakers are designed to provide consistent, uniform shaking action. The back-and-forth reciprocating motion has a 19 mm stroke length. Permanently lubricated ball bearings and maintenance-free, brushless DC motor provide reliable service and continuous-duty operation.
Catalog No. 50-222-9908
Explore available promotions