Learn More
Grainger GAUGE, O-RING SIZING
Supplier:  Grainger 5JKC0
Encompass_Preferred
The Grainger 5JKC0 O-Ring Sizing Gauge is a tool used to accurately measure the inside diameter of O-rings and other rubber seals. It features 30 precision-machined stainless steel pins in all standard O-ring sizes from 1/16" to 1-1/4" ID in 1/16" increments. The pins are marked with the O-ring ID size for easy sizing. This high quality gauge allows you to quickly identify the replacement O-ring needed by finding the best fit on the pin set. Made in the USA, it comes with a fitted plastic storage case for protection and organization. Overall an essential tool for any maintenance department working with hydraulic/pneumatic seals.
Catalog No. NC2282041
Explore available promotions