Learn More
Hach Company BOD Nutrient Buffer Pillows, 4 mL concentrate to make 4 L, 50/pk
Supplier: Hach Company 2436466
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
For preparation of BOD5 dilution water. Each pillow combines the APHA-specified buffer andnutrients to prepare 4 liters of dilution water. Pack of 50 pillows.