Learn More
STEMCELL Technologies Uridine, Size: 5 g
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: STEMCELL Technologies 1000540
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Uridine is one of the five nucleosides used for RNA synthesis and as a building block for other biomolecules (Tsukimoto). Uridine 5’-diphosphate and uridine 5’-triphosphate are signaling molecules that activate the purinergic receptors (Tsukimoto). Uridine is also used as a medium supplement for culturing primary hippocampal neuron cells (François-Moutal et al.).CANCER RESEARCH· Uridine has been used to reduce 5-fluorouracil toxicity, without affecting its antitumor activity (Pizzorno et al.).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.