Learn More
Cole-Parmer International Medcom RADALERT 100 Medcom Radalert 100X Nuclear Radiation Monitor
Supplier:  Cole-Parmer 5051103
Encompass_Preferred
Many police departments, fire departments, and other emergency response agencies as well as laboratories, and industries use the Radalert 100. The digital display shows readings in your choice of counts per minute (up to 100,000 cpm) or mR/hr (up to 100 mR/hr) or in accumulated counts. A red LED blinks and a beeper chirps with each count. An audible alert sounds when the radiation reaches a user-adjustable level.
Catalog No. 50-631-457
Explore available promotions