Learn More
Henke-Sass, Wolf Non-Sterile 2-Part Syringes
![SDP](/content/dam/fishersci/glyphs/sdp.png)
These syringes feature a 2-part construction containing no rubber and eliminating the need for silicone oil. Made from polypropylene and polyethylene.
Supplier: Henke-Sass, Wolf ABC30LS
Description
- Disposable Syringes
- 2-Part Syringe
- Unsterile
- Polypropylene Barrel
- Polyethylene Plunger
- Single-use
- Latex Free
- ISO
- Chromatography
- Laboratory Use
Specifications
Not Autoclavable | |
Polyethylene (Plunger), Polypropylene (Barrel) | |
50 Syringes | |
Yes | |
30 mL | |
Yes |
ISO 13485 | |
Clear | |
Yes | |
Bulk | |
Leur Slip | |
Non-sterile |