Learn More
Enzo Life Sciences nNOS (rat), (recombinant) (10µg)
Supplier: Enzo Life Sciences ALX2018540010
Alternative name: Nitric oxide synthase (neuronal), NOS I. Purity: ≥90% (SDS-PAGE). MW: ~160kDa/subunit; homodimer. Formulation: Liquid. In 50 mM TRIS, pH 7.4, containing 0.1 mM EDTA, 10% glycerol, 100 mM NaCl, 0.1 mM DTT, 10 µM tetrahydrobiopterin. Source: Produced in E. coli. Untagged rat nNOS. Long Term Storage: -80°C. Use/Stability: Stable for at least 1 year after receipt when stored at -80°C. Keep stock vial on ice during experiments. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -80°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.