Learn More
Nitinol foil, 0.05mm (0.002in) thick, superelastic, flat annealed, pickled surface, Thermo Scientific Chemicals
$88.20 - $930.50
Specifications
Chemical Name or Material | Nitinol foil |
---|---|
Name Note | 0.05mm (0.002 in.) thick, superelastic, flat annealed, pickled surface |
MDL Number | MFCD02091734 |
Solubility Information | Insoluble in water. |
Odor | Odorless |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AA45492FF
|
Thermo Scientific Chemicals
045492FF |
25 x 25 mm |
Each for $88.20
Order in Bulk
Request bulk or custom formats |
|
|||||
AA45492FI
|
Thermo Scientific Chemicals
045492FI |
50 x 50 mm |
Each for $289.50
Order in Bulk
Request bulk or custom formats |
|
|||||
AA45492GH
|
Thermo Scientific Chemicals
045492GH |
100 x 100 mm |
Each for $930.50
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
Nitinol foil is used in cell-phone technology as a retractable antenna, or microphone boom, due to its highly flexible and mechanical memory nature. It is also used in some novelty products, such as self-bending spoons which can be used by amateur and stage magicians to demonstrate psychic powers or as a practical joke, as the spoon will bend itself when used to stir tea, coffee, or any other warm liquid. It is highly biocompatible and has properties suitable for use in orthopaedic implants. Due to Nitinol's unique properties it has seen a large demand for use in less invasive medical devices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Nitinol foil | |
MFCD02091734 | |
Odorless |
0.05mm (0.002 in.) thick, superelastic, flat annealed, pickled surface | |
Insoluble in water. | |
≈0.25g/25x25mm |
Safety and Handling
GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.
GHS P Statement
P260-P261-P280-P363-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.