Learn More
Perkin Elmer US LLC NIR Quartz SUPRASIL Rectangular Semi-Micro Cell with Stopper - 10 mm, Qty. 2
Supplier: Perkin Elmer US LLC B0631066
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The NIR Quartz SUPRASIL 300 Semi-Micro cells are widely used in routine UV/Vis liquids analysis. They have the same outside dimensions as Macro cells, but their cell walls have been thickened slightly, limiting the interior sample chamber width to 4 mm with a base thickness of 3.2 mm. Each cell has two clear windows. Semi-Micro cells have either a lid or a stopper. While the outside dimensions of Semi-Micro cells with Stoppers differ slightly from those with Lids, the path length and volume are not affected. NIR Quartz SUPRASIL 300 is a synthetic Quartz free from OH absorption. An empty SUPRASIL 300 cell gives a transmission of more that 80% over a spectral range of 200 - 3,500 nm. These cells are identified by a red PerkinElmer logo.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.