Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
β-Nicotinamide Adenine Dinucleotide, MP Biomedicals™
Click to view available options
Quantity:
100 mg
500 mg
5 g
10 g
Description
β-NAD occurs in living cells primarily in the oxidized state.
- Serves as a coenzyme of hydride-transfer enzymes in dehydrogenases, especially in the dehydrogenation of primary and secondary alcohols
- Usually acts as a hydrogen acceptor, forming NADH, which then serves as a hydrogen donor in the respiratory chain
- Used as a substrate for NAD(+)-consuming enzymes, which include ADP-ribose transferases, poly(ADP-ribose) polymerases, cADP-ribose synthases and sirtuins
- Recently reported to be a P2Y11 agonist
Specifications
Absorbance | λ max (water): 259 ± 3nm |
Melting Point | 160°C (literature) |
pH | 2 to 4 (1% aq. soln.) |
Quantity | 500 mg |
Solubility Information | Soluble in water (200mg/mL) |
Formula Weight | 663.4 (anhydrous) |
Percent Purity | ≥98% |
Physical Form | Powder |
Safety and Handling
Recommended Storage | 0°C, desiccate |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction