Learn More
Glass Expansion Nickel Skimmer Hot & Cold Plasma Thermo Q/RQ/TQ
Supplier: Glass Expansion TG1035NI
Nickel Skimmer Cone, Cold and Hot Plasma (1341420) – Suitable for Thermo Q/RQ/TQ ICP-MS. Specifically designed to operate ONLY with the Cold Plasma Lens Kit (1341380). This skimmer cone can be used with the Cold Plasma Lens Kit to operate in both Cold (RF Power 550 W) and Hot Plasma conditions (RF Power 1550 W). This reduced plasma intensity configuration is typically used to suppress Ar interferences for elements, such as K and Ca; and/or for the analysis of other light elements like Li and Na. Not for samples containing HF. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. Use Skimmer ConeGuard (70-803-1028) to protect cone threads during cleaning. When not in use, keep cones stored in original packaging for protection.For high matrix tolerance and high sensitivity, we recommend NC2046959. For Robust plasma conditions, we recommend TG1045-Ni.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.