Learn More
Glass Expansion Nickel Sampler Cone, Wet Plasma (319-645) For Nu Instruments Plasma2, Plasma3, Attom ES and Vitesse TOF-ICP-MS. Suitable for standard applications.
Supplier: Glass Expansion NU1011ANI
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Nickel Sampler Cone, Wet Plasma (319-645) For Nu Instruments Plasma2, Plasma3, Attom ES and Vitesse TOF-ICP-MS. Suitable for standard applications. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.