Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nickel Aluminum, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
Description
Nickel Aluminum is used as wear resistant coatings. It is also used for the preparation of Raney™ Nickel catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Black to Gray |
Avg. Mol. Wt. or Mol. Wt. Range | Ni:Al; 50:50 wt% |
Quantity | 500 g |
UN Number | UN3208 |
Solubility Information | Insoluble in water. |
Odor | Odorless |
Sensitivity | Water reactive |
Form | Powder |
Chemical Name or Material | Nickel Aluminum |
Safety and Handling
Hazard Category | H261-H350 |
Hazard Statement | GHS H Statement H228-H261-H372-H351-H317 Flammable solid. In contact with water releases flammable gas. Causes damage to organs through prolonged or repeated exposure. Suspected of causing cancer. May cause an allergic skin reaction. |
Precautionary Statement | P201-P202-P223-P231+P232-P281-P308+P313-P335+P334-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: METALLIC SUBSTANCE, WATER-REACTIVE, N.O.S. |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction