Learn More
Rocky Mountain Biologicals, LLC Newborn Bovine Calf Serum (NBS) U.S. Origin 500ml
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Rocky Mountain Biologicals, LLC NBSBBT5XM
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
RMBIO’s Newborn Bovine Calf Serum (NBS) is a high-quality supplement for use in a variety of cell culture media. NBS can serve as a low-cost alternative to FBS while providing many of the same nutrients. NBS is manufactured and processed according to the highest standards and is carefully monitored to ensure product safety and performance. The average endotoxin level is < 2 EU/ml and the average hemoglobin level is < 20 mg/dl. All lots of NBS are of US origin, are 9CFR tested, and 0.2 µm filtered. Product should be stored at ≤ -10° C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.