Learn More
R & D SYSTEMS INC Newborn Calf Serum, Heat Inactivated
SDP
Supplier:  R & D SYSTEMS INC S11295H
Encompass_Preferred
Call your Customer Service Representative for Bulk Discounts. R&D Systems Newborn Calf Serum (NBC, NBCS) provides many of the same nutrients as Fetal Bovine Serum, but has slightly higher protein levels. For routine maintenance of select cell lines and where low antibody concentrations are not critical, Newborn Calf Serum is often used as a lower-priced alternative to FBS. Heat Inactivated Newborn Calf Serum is treated by heat inactivation where serum is exposed to a temperature of 56 °C for 30 minutes under controlled conditions. All R&D Systems Sera are manufactured in an ISO 9001:2015 certified facility.
Catalog No. 50-152-7075
Explore available promotions