Learn More
LABSYSTEMS DIAGNOSTICS OY Biotinidase activity from DBS
Supplier: LABSYSTEMS DIAGNOSTICS OY 6199821
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Neonatal Biotinidase kit is for enzymatic determination of human biotinidase enzyme activity from blood samples dried on filter paper. Intended for newborn screening of biotinidase deficiency. The kit can be used for both manual and automate use