Learn More
Fujifilm Healthcare Americas Corp NEFA Linearity Material
Supplier:  Fujifilm Healthcare Americas Corp 99776491
Encompass_Preferred
The NEFA Linearity Material is for use with the HR Series NEFA-HR (2) assay for the determination of linearity and/or calibration verification of non-esterified fatty acid analyte. The HR Series NEFA HR (2) is an in vitro enzymatic colorimetric method assay for the quantitative determination of non-esterified fatty acids (NEFA) in serum. The NEFA Linearity Set is a solution of 2.5 mEq/L Oleic Acid. Package size: 1 x10 mL bottle. This product is For In Vitro Diagnostic Use. Contact supplier for additional information at www.ivd.fujimed.com.
Catalog No. NC9349895