Learn More
World Precision Instrument Needle Microprobes, High Temperature-23ga, 8cm (MT-23/8HT)
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: World Precision Instrument MT23/8HT
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
When precise temperature measurements are required, WPI can provide you with a very accurate monitor and thermocouple microprobes. WPI monitors have both resolution and accuracy of 0.1°C in the 0-50°C range and are traceable to NIST standards, whereas, other competitive electronic thermometers have an accuracy that is usually to 0.5°C or worse. Furthermore, all our type T clinical probes are guaranteed accurate to 0.1°C, due to our stringent wire standards. These are five times more accurate than competitive probes made with regular “Special Limits” wire.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.