Learn More
Glass Expansion Nebulizer Hose Clip
Supplier: Glass Expansion SNP1
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Nebulizer Pressure Hose Clips. Package of 10 plastic nebulizer argon supply hose clamps. Used to secure a soft walled argon supply tubing to the EzyLok 6mm Hose Adaptor (70-808-0735) using the EzyLok Kit (EL-1).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001