Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ NAV 26
Selective Nav1.7 channel blocker
$276.08 - $1171.60
Specifications
Purity | >98% |
---|---|
Product Type | NAV 26 |
Molecular Weight (g/mol) | 434.41 |
Inhibitors | NAV 26 |
Description
Selective Nav1.7 channel blocker (IC50 = 370 nM). Exhibits >85-fold selectivity for Nav1.7 over Nav1.5 and hERG channels. Reduces nociceptive behavior in rat models of pain.Specifications
>98% | |
NAV 26 | |
434.41 | |
NAV 26 |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title