Learn More
Yamato Scientific America, Inc. Yamato 28L Natural Convection Oven, 220V
Supplier: Yamato Scientific America, Inc. DX312C
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Standard 28L natural convection constant temperature drying oven with extensive features and simple operation. Economical and cost saving. Easy to use and maintain. Excellent temperature accuracy. Digital PID controller, calibration off-set and increased safety and self diagnostic functions. Operating temp ambient +5~300°C