Learn More
CTL Scientific Supply Corp NANOCOLOR Phenolic index 5, contains 20 tube tests
Supplier: CTL Scientific Supply Corp 985074
Tube test for the determination of Phenols. Precise rapid tests for all kind of water and waste water samples. Time-saving and reliable analysis together with Macherey-Nagel NANOCOLOR photometers. Measuring range: Phenols - 0.2–5.0 mg/L PhenolManufactured by Macherey-Nagel