Learn More
Proteintech Group Inc Nano-Secondary alpaca anti-mouse IgG2b, recombinant VHH, for 2x Cys conjugation [CTK0106]
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Proteintech Group Inc SMSG2BCYS22500
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Monovalent, recombinant secondary single domain antibody to mouse IgG2b: alpaca monoclonal Nanobody, Fc-specific, for cysteine conjugation with thiol-reactive reagents, e.g. maleimides
This item is not returnable.
View return policy