Learn More
Beckman Coulter Multisizer™ 4e Smart Technology Aperture Tubes
Smart Technology Aperture Tubes are designed specifically for the Multisizer 4 or 4e Analyzer. The aperture allows extended sizing measurement range of 2% to 80% of the aperture diameter.
Supplier:  Beckman Coulter A36401
 View more versions of this product
Catalog No. 15-655-150
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
  • Extended measurement range (2% to 80% of aperture diameter) leads to less frequent aperture changes for more analysis options
  • A wide range of apertures is available with diameters ranging from 10 to 2,000 μm
  • Specifically designed for the Multisizer 4e or Multisizer 4 Coulter Counters
  • 100 μm ST Aperture Tube is standard with the Multisizer 4e Coulter Counter
  • Compatible with ST Beakers and Accuvette STs
  • Convenient tubes ensure accurate, and consistent results
  • Size and serial number information is included on a bar code on the tube for automatic data entry via instrument bar code reader
Specifications
Specifications
Aperature Tube
40 μm to 1600 μm
Multisizer™ 4 or 4e Coulter Counters
Glass
2000μm
Product Suggestions
Product Suggestions