Learn More
Electron Microscopy Sciences SEM Multi Pin Stub Holder for 12 Hitachi M4
Supplier: Electron Microscopy Sciences 7595212H
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
This 62 mm in diameter multi pin holder is designed to save time. It is made from machined aluminum with stainless steel Allen wrench screws. It accommodates up to twelve standard 12.7 mm (1/2") pin stubs
Type: Hitachi M4