Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Analog Variable-Speed Console Drive, 7 to 200 rpm; 115 VAC
Supplier: AVANTOR MASTERFLEX (VWR LLC) MFLX0755510
Drives accept nine different pump head types: Standard, Easy-Load, Easy-Load II, Easy-Load 3, Multichannel, High-Performance, Cartridge, PTFE tubing, and PTFE-diaphragm. Drives 07555-00 and -05 are not recommended for use with the PTFE-tubing and PTFE-diaphragm pump heads.