Learn More
MSA™ Ultravue™ Full Facepiece Assembly
Facepiece without nosecup has inhalation check valve and exhalation valve that work together to avoid exhaled air from entering the mask mounted regulator and contaminating the internal regulator components.
Supplier: MSA™ 801542
Safety Services Available
Overview
Repair/Recertification
This item is not returnable.
View return policy
Description
- Pressure-Demand Ultravue
- Full Facepieces
- Rubber hycar material
- Standard 5-point head harness
- Without nosecup
Specifications
NIOSH | |
Facepiece Assembly | |
Hycar, MMR |
Pressure-Demand | |
Quick-Disconnect | |
Medium |