Learn More
MSA™ Advantage™ 3100 Series Gas Mask Replacement Canister
Supplier: MSA™ 10059903
This item is not returnable.
View return policy
Description
The small, round plastic canister attaches directly to the facepiece
- Canisters are seamless to prevent leakage or channeling of gases/vapors past the chemical sorbents
- Sealing tape on the bottom air intake keeps them fresh and prevents moisture from entering the canister before use
- Compact oval shape for easy use and storage
- Labeled as to type and limitations
- Approved for Phosphine/Ammonia/Chlorine/Oil Based Particulates (PH/AM/CL/P100)
- Color-coded olive/magenta for fast and accurate recognition
- NIOSH Approval TC-14G-0274