Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IgG1, Alexa Fluor 647, Isotype Control, R&D Systems™
Mouse Monoclonal Antibody has been used in 1 publication
Supplier: R&D Systems IC002R
This item is not returnable.
View return policy
Description
IgG1 Monoclonal specifically detects IgG1 in Mouse samples. It is validated for Control.Specifications
IgG1 | |
11711 | |
Control 5 uL/10^6 cells | |
IgG1, Igh-4, RGD1359539, VH7183 | |
KLH | |
200 Tests | |
Secondary | |
Mouse | |
Supernatant |
Monoclonal | |
Alexa Fluor 647 | |
Supplied in a saline solution containing BSA and Sodium Azide. | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Selected for use as mouse IgG 1 isotype control for flow cytometry applications. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 κ |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction