Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Molecular Sieve Dehydrate with Indicator For Drying Gases, Beads, Honeywell Fluka™
Beads, with indicator for drying gases
$875.00
Specifications
Density | 2g/cm |
---|---|
Pore Size | 4 |
Packaging | Glass Bottle |
Chemical Name or Material | Molecular Sieve Dehydrate Fluka With Indicator For Drying Gases |
CAS | 1318-02-1, 10124-43-3 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
60-048-492
|
Honeywell Chemicals
282111KG |
1 kg | Glass Bottle |
Each of 1 for $875.00
|
|
||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
2g/cm | |
Glass Bottle | |
1318-02-1, 10124-43-3 |
4 | |
Molecular Sieve Dehydrate Fluka With Indicator For Drying Gases | |
NONH for all modes of transport |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title