Learn More
Thermo Scientific MODEL 5020 GUARD CELL
Supplier: Thermo Scientific 550417
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Guard Cell, CoulometricModel 5020 for use with Coulochem detectors - designed for inline removal of electroactive impurities in mobile phases for HPLC-ECD, rated to 41.3 Mpa (6000 psi)
Description
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)