Learn More
Sentry Air Systems Model 200 Dual Arm Fume Extractor
Supplier: Sentry Air Systems SS200FSD
The Dual Arm Fume Extractor provides source capture fume extraction for two operators simultaneously. The dual 52 in. (132.08 cm) self-supportive flex arms combined with a powerful fan direct fumes and particulate away from the operators breathing zone into the filtration system. This unit can be mounted on the wall or bench, or placed on the floor to save space. The Dual Arm Fume Extractor offers fume extraction for a variety of applications including chemical fumes, solvent and epoxy fumes, dust, and other lab processes. Approximate Airflow: up to 80 CFM (40 per arm). Filters Available: HEPA Filter [up to 99.97% efficiency on particles 0.3 microns and larger], ASHRAE Filter [up to 95% efficiency on particles 0.5 microns and larger], ULPA Filter [up to 99.995% efficiency on particles 0.13 microns and larger], Activated Carbon, and specialty-blended filtration [i.e. acid gas, mercury, aldehyde, and ammonia].
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.