Learn More
Sentry Air Systems Model 200 Portable Fume Extractor
Supplier: Sentry Air Systems SS200FS
The Model 200 Portable Fume Extractor uses powerful suction and 43 in. (109.22 cm) long self-supportive flex arm to source capture fumes and particulate. The Floor Sentry sits on the floor saving valuable benchtop space. The self-supportive flex arm can easily be maneuvered and placed close to the fume or particulate emitting procedure. The Portable Fume Extractor directs fumes through the capture hose into the filtration system and releases the filtered air back into the surrounding room. This compact and portable system provides a fume control solution for chemical fumes, solvent and epoxy fumes, and other processes that produce fumes and/or particulate. Approximate Airflow: up to 80 CFM. Filters Available: HEPA Filter, ASHRAE Filter, ULPA Filter, Activated Carbon, and specialty-blended filtration [i.e. acid gas, mercury, aldehyde, and ammonia].
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.