Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ML 154

Potent neuropeptide S receptor (NPSR) antagonist

Specifications

Purity >99%
Product Type ML 154
Molecular Weight (g/mol) 545.47
Inhibitors ML 154
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
51-611-0
View Documents
Tocris Bioscience™
5161/10
10 mg Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
51-615-0
View Documents
Tocris Bioscience™
5161/50
50 mg Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
Description

Description

Potent neuropeptide S receptor (NPSR) antagonist (pA2 = 9.98). Inhibits neuropeptide S-induced ERK phosphorylation over cAMP responses and calcium responses (IC50 values are 9.3, 22.1 and 36.5 nM, respectively). Appears to modulate addictive behavior in vivo. Displays no activity against vasopressin V1B receptors. Brain penetrant.
Specifications

Specifications

>99%
ML 154
545.47
ML 154
SDS
Documents

Documents

Product Certifications

For Research Use Only

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.