Learn More
GUARDIAN EQUIPMENT Thermostatic Mixing Valve, 50 Gallon/189 Liter Capacity
Supplier: GUARDIAN EQUIPMENT G6040
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Thermostatic mixing valve to blend hot and cold water to deliver tepid water. Valve has flow capacity of 50 gallons (189 liters) per minute at 30 PSI (2.1 bar) pressure drop. Valve can be used to supply emergency shower or combination safety station. Depending on water supply size and pressure, valve can supply multiple units.