Learn More
Chemglass Life Sciences Mistral Sample Concentrator, Without Needle Head, Porvair, 220V
SDP
Supplier:  Chemglass Life Sciences CLS3599C100E
Encompass
Request Bulk Order
Mistral Sample Concentrator, Without Needle Head, Porvair, 220V. The Ultravap Mistral from Porvair Sciences is designed to remove the traditional laboratory ‘bottleneck’ of solvent evaporation from microplates. Fully automating the dry-down step has always been impossible because it is difficult to interface liquid handling robots with traditional centrifugal-type evaporators. The Ultravap Mistral design, by dispensing with the rotating arm of the centrifuge, overcomes this problem and is much better positioned to link with your robot. With more than 20 years’ experience in producing deep well microplates, Porvair Sciences has thoroughly researched the problems of drying down organic solvents in plates. This has led to the ultimate microplate blow down evaporator – the Ultravap Mistral.
Catalog No. 50-194-1140