Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Minigrip™ Premium Red Line™ Reclosable Zipper Bags
Many size and film thickness options
Supplier: Minigrip™ MGRL2P0303
Description
- Premium zipper easily snaps open and zips securely shut
- Superior clarity that you can see
- Most sizes are packaged in convenient DispenserBag™ packaging
- Packaged in high quality corrugated boxes for reliable storage and easy handling
- Manufactured with 100% FDA-approved virgin LDPE and LLDPE resins
- Reinforced zipper and side seams to keep bags from splitting when opened or filled
Specifications
Zip Seal | |
LDPE, LLDPE | |
3 in. |
Clear | |
2 mil | |
3 in. |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction