Learn More
Research Products International Corp Minidizer Hybridization Oven
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Research Products International Corp 95033001
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Compact size is excellent for limited bench space. Rotation speed is set at 12 rpm with precision temperature controls. Has a bottle capacity of four 35 x 150mm bottles, four 50 ml and eight 15 ml conical plastic bottles. Minidizer has consistent, repeatable results achieved with microprocessor controls. The chamber interior is stainless steel fabrication with a 3/8 inch beta blocking acrylic cover. Cover swings down for easy access to the bottle carousel. Carousel pops out of chamber for easy insertion or removal of bottles.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.