Learn More
Research Products International Corp Mini-Blotting Containers, 9.0 x 6.4 x 2.1cm, Black, 6 per Case
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Research Products International Corp 248716B
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Plastic containers for many laboratory applications. Containers have a smooth, flat surface that is ideal for blotting membranes or staining gels. Hinged lid with snap lock closure prevents evaporation of reagents. Constructed of ultra-clear polystyrene.