Learn More
World Precision Instrument Mini Centrifuge, blue lid, 2 rotors, 110V
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: World Precision Instrument 504321
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
This personal centrifuge fits in the palm of your hand. It is deal for quick spin downs of either microtubes or PCR tubes. Close the lid and the rotor instantly ramps up to 6000 RPM. Open the lid and it decelerates to a stop. The 8-position microtube rotor is incuded, along with a second rotor for 0.2mL PCR tubes and strips. The second rotor is stored (with the removal key) in a compartment on the bottom of the centrifuge.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.