Learn More
Perkin Elmer US LLC Mid Infrared Polystyrene TRM for ATR Accessories
Supplier: Perkin Elmer US LLC L1600133
The TRM is traceable to a National Institute of Standards and Technology (NIST) Standard Reference Material (SRM 1921b), obtained from NIST, Gaithersburg, Maryland, MD20899, USA. The traceability path is highlighted on the TRM certificate provided with the sample.