Learn More
World Precision Instrument MicroTip - Fiber Optic Oxygen Sensor-501656-C
Supplier: World Precision Instrument 501656C
The MicroTip is a needle-type (27 ga.) oxygen micro sensor designed for applications where a small tip size of 50 µm and a fast response time (t90) of 1 s are necessary. The oxygen sensitive sensor tip consists of 140 µm fiber tapered to a 50 µm tip. The sensor is housed inside a stainless steel needle of 22 mm length and 0.4 mm diameter. This allows penetration through a septum rubber or similar material. These sensors are ideal for oxygen profiling in sediments and biofilms.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.