Learn More
Research Products International Corp SuperClear MicroTubes, 2000ul, RNBW, 500/cs
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Research Products International Corp 145431LC
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
SuperClear MicroTubes complement all current molecular biology applications. Tubes are manufactured from optically clear polypropylene resin (USP class IV) that allows clear view of contents and will not interfere with your ability to detect DNA or PCR. Extra thick and durable wall withstands centrifugation to 20,000 x G for exceptional durability.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.